Please login to your online account to display your discounted pricing

anti-Bromodeoxyuridine/BrdU, Clone: BU20a, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP232922

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP232922

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity All
Immunogen Bromodeoxyuridine (BrdU) conjugated to KLH
Clone BU20a
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Quantity 0.5mL
Isotype IgG1
Antigen Bromodeoxyuridine/BrdU
Regulatory Status RUO
Format Purified
Formulation PBS with 0.05% BSA.
Conjugate Unlabeled
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Concentration 0.2mg/mL
Host Species Mouse