Please login to your online account to display your discounted pricing

anti-Bromodeoxyuridine/BrdU, Clone: BU20a, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone BU20a
Host Species Mouse
Isotype IgG1
Immunogen Bromodeoxyuridine (BrdU) conjugated to KLH
Antigen Bromodeoxyuridine/BrdU
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP2329221M

 
novus biologicals canada
NBP2329221ML
Unlabeled 1mL Each for $429.00

NBP232922

 
novus biologicals canada
NBP232922
Unlabeled 0.5mL Each for $321.00
Description & Specifications

Specifications

Clone BU20a
Host Species Mouse
Isotype IgG1
Immunogen Bromodeoxyuridine (BrdU) conjugated to KLH
Antigen Bromodeoxyuridine/BrdU
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL