Please login to your online account to display your discounted pricing

anti-Bromodeoxyuridine/BrdU, Clone: BRD494, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP2294141ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP2294141M

  • / Each
 

Description & Specifications

Specifications

Format Concentrated
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Quantity 1mL
Immunogen A recombinant fragment specific to human BrdU protein was used as the immunogen for the BRD494 antibody.
Isotype κ
Cross Reactivity All
Antigen Bromodeoxyuridine/BrdU
Host Species Mouse
Clone BRD494
Formulation Culture Medium
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Test Specificity It reacts with Bromodeoxyuridine (BrdU) in single stranded DNA (produced by partial denaturation of double stranded DNA), BrdU coupled to a protein carrier, as well as free BrdU. BrdU is a thymidine analog, incorporated into cell nuclei during DNA synthesis prior to mitosis. Antibody to BrdU is helpful in detecting S-phase cells, providing useful information on the aggressiveness of tumors.
Conjugate Unlabeled