Please login to your online account to display your discounted pricing

anti-Bromodeoxyuridine/BrdU, Clone: BRD494, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP229414

 View all options for this product


For Research Use Only

Catalog No. NBP229414


Description & Specifications


Antigen Bromodeoxyuridine/BrdU
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Clone BRD494
Conjugate Unlabeled
Cross Reactivity All
Format Concentrated
Formulation Culture Medium
Host Species Mouse
Immunogen A recombinant fragment specific to human BrdU protein was used as the immunogen for the BRD494 antibody.
Isotype κ
Quantity 0.5mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Test Specificity It reacts with Bromodeoxyuridine (BrdU) in single stranded DNA (produced by partial denaturation of double stranded DNA), BrdU coupled to a protein carrier, as well as free BrdU. BrdU is a thymidine analog, incorporated into cell nuclei during DNA synthesis prior to mitosis. Antibody to BrdU is helpful in detecting S-phase cells, providing useful information on the aggressiveness of tumors.