Please login to your online account to display your discounted pricing

anti-Bromodeoxyuridine/BrdU, Clone: BRD494, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone BRD494
Host Species Mouse
Isotype κ
Immunogen A recombinant fragment specific to human BrdU protein was used as the immunogen for the BRD494 antibody.
Antigen Bromodeoxyuridine/BrdU
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP229414

 
novus biologicals canada
NBP229414
Unlabeled 0.5mL Each for $321.00

NBP2294141M

 
novus biologicals canada
NBP2294141ML
Unlabeled 1mL Each for $429.00
Description & Specifications

Specifications

Clone BRD494
Host Species Mouse
Isotype κ
Immunogen A recombinant fragment specific to human BrdU protein was used as the immunogen for the BRD494 antibody.
Antigen Bromodeoxyuridine/BrdU
Monoclonal or Polyclonal Monoclonal
Format Concentrated
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Test Specificity It reacts with Bromodeoxyuridine (BrdU) in single stranded DNA (produced by partial denaturation of double stranded DNA), BrdU coupled to a protein carrier, as well as free BrdU. BrdU is a thymidine analog, incorporated into cell nuclei during DNA synthesis prior to mitosis. Antibody to BrdU is helpful in detecting S-phase cells, providing useful information on the aggressiveness of tumors.
Formulation Culture Medium