Please login to your online account to display your discounted pricing

anti-Bromodeoxyuridine/BrdU, Clone: BRD469, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP2329181ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP2329181M

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Host Species Mouse
Cross Reactivity All
Immunogen Bromodeoxyuridine (BrdU) conjugated to KLH
Format Purified
Clone BRD469
Isotype IgG1
Antigen Bromodeoxyuridine/BrdU
Quantity 1mL
Conjugate Unlabeled
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Regulatory Status RUO