Please login to your online account to display your discounted pricing

anti-Bromodeoxyuridine/BrdU, Clone: BRD469, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP232918

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP232918

  • / Each
 

Description & Specifications

Specifications

Antigen Bromodeoxyuridine/BrdU
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Clone BRD469
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity All
Format Purified
Formulation PBS with 0.05% BSA.
Host Species Mouse
Immunogen Bromodeoxyuridine (BrdU) conjugated to KLH
Isotype IgG1
Quantity 0.5mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal