Please login to your online account to display your discounted pricing

anti-BRIP1/FANCJ, Clone: pp15-IB4, Novus Biologicals™

$109.00 - $309.05

Specifications

Clone pp15-IB4
Host Species Murine
Isotype κ
Immunogen A peptide corresponding to the C-terminus of human FANCJ (within amino acids 1200-1249). [SwissProt No. Q9BX63]
Antigen BRIP1/FANCJ
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB100189SS

 
novus biologicals canada
NB100189SS
Unlabeled 0.025mL Each for $109.00

NB100189

 
novus biologicals canada
NB100189
Unlabeled 0.1mL Each for $309.05
Description & Specifications

Specifications

Clone pp15-IB4
Host Species Murine
Isotype κ
Immunogen A peptide corresponding to the C-terminus of human FANCJ (within amino acids 1200-1249). [SwissProt No. Q9BX63]
Antigen BRIP1/FANCJ
Monoclonal or Polyclonal Monoclonal
Format Ascites
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunoprecipitation
Applications Western Blot
Gene Accession No. Q9BX63
Gene ID (Entrez) 83990