Please login to your online account to display your discounted pricing

anti-BRIP1/FANCJ, Clone: GO11-3E6, Novus Biologicals™


Manufacturer: novus biologicals canada  NB100350SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB100350SS

  • / Each
 

Description & Specifications

Specifications

Antigen BRIP1/FANCJ
Applications Immunoprecipitation
Applications Western Blot
Clone GO11-3E6
Conjugate Unlabeled
Cross Reactivity Human
Format Ascites
Gene Accession No. Q9BX63
Host Species Murine
Immunogen Fusion protein corresponding to amino acids 647-1043 of human FANCJ. [Swiss-Prot No. Q9BX63]
Isotype κ
Quantity 0.025mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 83990
Target Species Human