Please login to your online account to display your discounted pricing

anti-BRIP1/FANCJ, Clone: GO11-3E6, Novus Biologicals™


Manufacturer: novus biologicals canada  NB100350

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB100350

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. Q9BX63
Primary or Secondary Primary
Conjugate Unlabeled
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 83990
Applications Immunoprecipitation
Applications Western Blotting
Isotype κ
Quantity 0.1mL
Format Ascites
Immunogen Fusion protein corresponding to amino acids 647-1043 of human FANCJ. [Swiss-Prot No. Q9BX63]
Antigen BRIP1/FANCJ
Regulatory Status RUO
Host Species Mouse
Clone GO11-3E6
Cross Reactivity Human