Please login to your online account to display your discounted pricing

anti-BRIP1/FANCJ, Clone: GO11-3E6, Novus Biologicals™

$109.00 - $309.05

Specifications

Clone GO11-3E6
Host Species Murine
Isotype κ
Immunogen Fusion protein corresponding to amino acids 647-1043 of human FANCJ. [Swiss-Prot No. Q9BX63]
Antigen BRIP1/FANCJ
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB100350

 
novus biologicals canada
NB100350
Unlabeled 0.1mL Each for $309.05

NB100350SS

 
novus biologicals canada
NB100350SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Clone GO11-3E6
Host Species Murine
Isotype κ
Immunogen Fusion protein corresponding to amino acids 647-1043 of human FANCJ. [Swiss-Prot No. Q9BX63]
Antigen BRIP1/FANCJ
Monoclonal or Polyclonal Monoclonal
Format Ascites
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunoprecipitation
Applications Western Blot
Gene Accession No. Q9BX63
Gene ID (Entrez) 83990