Please login to your online account to display your discounted pricing

anti-BRCA1, Clone: RAY, Novus Biologicals™


Manufacturer: novus biologicals canada  NB100598

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB100598

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Isotype IgG2a
Clone RAY
Format Purified
Immunogen Human BRCA1 corresponding to residues 1314-1864 [UniProt No. P38398]
Regulatory Status RUO
Antigen BRCA1
Formulation PBS
Monoclonal or Polyclonal Monoclonal
Gene Accession No. P38398
Primary or Secondary Primary
Concentration 2.76mg/mL
Quantity 0.1mL
Gene ID (Entrez) 672
Conjugate Unlabeled
Host Species Mouse
Cross Reactivity Human