Please login to your online account to display your discounted pricing

anti-BRCA1, Clone: RAY, Novus Biologicals™


Manufacturer: novus biologicals canada  NB100598SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB100598SS

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 672
Formulation PBS
Monoclonal or Polyclonal Monoclonal
Gene Accession No. P38398
Concentration 2.76mg/mL
Primary or Secondary Primary
Format Purified
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Quantity 0.025mL
Isotype IgG2a
Antigen BRCA1
Clone RAY
Cross Reactivity Human
Immunogen Human BRCA1 corresponding to residues 1314-1864 [UniProt No. P38398]
Host Species Mouse
Regulatory Status RUO