Please login to your online account to display your discounted pricing

anti-BRCA1, Clone: MU, Novus Biologicals™


Manufacturer: novus biologicals canada  NB100600

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB100600

  • / Each
 

Description & Specifications

Specifications

Formulation Tris-glycine, 150mM NaCl
Gene Accession No. P38398
Primary or Secondary Primary
Gene ID (Entrez) 672
Concentration 1.9mg/mL
Conjugate Unlabeled
Antigen BRCA1
Host Species Mouse
Monoclonal or Polyclonal Monoclonal
Format Purified
Clone MU
Isotype κ
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Human BRCA1 corresponding to residues 1314-1864 [UniProt No. P38398]
Quantity 0.1mL
Cross Reactivity Human
Regulatory Status RUO