Please login to your online account to display your discounted pricing

anti-BRCA1, Clone: MU, Novus Biologicals™

$109.00 - $329.00

Specifications

Clone MU
Host Species Mouse
Isotype κ
Immunogen Human BRCA1 corresponding to residues 1314-1864 [UniProt No. P38398]
Antigen BRCA1
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB100600

 
novus biologicals canada
NB100600
Unlabeled 0.1mL Each for $329.00

NB100600SS

 
novus biologicals canada
NB100600SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Clone MU
Host Species Mouse
Isotype κ
Immunogen Human BRCA1 corresponding to residues 1314-1864 [UniProt No. P38398]
Antigen BRCA1
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Gene Accession No. P38398
Gene ID (Entrez) 672
Formulation Tris-glycine, 150mM NaCl
Concentration 1.9mg/mL