Please login to your online account to display your discounted pricing

anti-BRCA1, Clone: MU, Novus Biologicals™


Manufacturer: novus biologicals canada  NB100600SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB100600SS

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. P38398
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Concentration 1.9mg/mL
Formulation Tris-glycine, 150mM NaCl
Conjugate Unlabeled
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Gene ID (Entrez) 672
Quantity 0.025mL
Format Purified
Host Species Mouse
Clone MU
Antigen BRCA1
Isotype κ
Immunogen Human BRCA1 corresponding to residues 1314-1864 [UniProt No. P38398]
Cross Reactivity Human
Regulatory Status RUO