Please login to your online account to display your discounted pricing

anti-BRCA1, Clone: KEN, Novus Biologicals™


Manufacturer: novus biologicals canada  NB100599SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB100599SS

  • / Each
 

Description & Specifications

Specifications

Antigen BRCA1
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blot
Clone KEN
Concentration 1.67mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation Tris-glycine, 150mM NaCl
Gene Accession No. P38398
Host Species Murine
Immunogen Human BRCA1 corresponding to residues 1314-1864 [UniProt No. P38398]
Isotype κ
Quantity 0.025mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 672
Target Species Human