Please login to your online account to display your discounted pricing

anti-BRCA1, Clone: KEN, Novus Biologicals™


Manufacturer: novus biologicals canada  NB100599

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB100599

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. P38398
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Concentration 1.67mg/mL
Formulation Tris-glycine, 150mM NaCl
Gene ID (Entrez) 672
Conjugate Unlabeled
Host Species Mouse
Isotype κ
Cross Reactivity Human
Format Purified
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Clone KEN
Antigen BRCA1
Quantity 0.1mL
Immunogen Human BRCA1 corresponding to residues 1314-1864 [UniProt No. P38398]
Regulatory Status RUO