Please login to your online account to display your discounted pricing

anti-Biotin (Vitamin B7 or Vitamin H), Clone: Hyb-8, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP232811

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP232811

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Cross Reactivity All
Quantity 0.5mL
Format Purified
Immunogen Biotinylated sheep immunoglobulin
Host Species Mouse
Clone Hyb-8
Antigen Biotin (Vitamin B7 or Vitamin H)
Concentration 0.2mg/mL
Conjugate Unlabeled
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Regulatory Status RUO
Primary or Secondary Primary
Isotype IgG1