Please login to your online account to display your discounted pricing

anti-Biotin (Vitamin B7 or Vitamin H), Clone: Hyb-8, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone Hyb-8
Host Species Mouse
Isotype IgG1
Immunogen Biotinylated sheep immunoglobulin
Antigen Biotin (Vitamin B7 or Vitamin H)
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP2328111M

 
novus biologicals canada
NBP2328111ML
Unlabeled 1mL Each for $429.00

NBP232811

 
novus biologicals canada
NBP232811
Unlabeled 0.5mL Each for $321.00
Description & Specifications

Specifications

Clone Hyb-8
Host Species Mouse
Isotype IgG1
Immunogen Biotinylated sheep immunoglobulin
Antigen Biotin (Vitamin B7 or Vitamin H)
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL