Please login to your online account to display your discounted pricing

anti-Biotin (Vitamin B7 or Vitamin H), Clone: Hyb-8, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP2328111ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP2328111M

  • / Each
 

Description & Specifications

Specifications

Antigen Biotin (Vitamin B7 or Vitamin H)
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Host Species Mouse
Immunogen Biotinylated sheep immunoglobulin
Isotype IgG1
Cross Reactivity All
Regulatory Status RUO
Quantity 1mL
Format Purified
Monoclonal or Polyclonal Monoclonal
Clone Hyb-8
Conjugate Unlabeled
Primary or Secondary Primary
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL