Please login to your online account to display your discounted pricing

anti-beta-III Tubulin, Clone: 2E9, Novus Biologicals™

$109.00 - $309.05

Specifications

Clone 2.00E+09
Host Species Mouse
Isotype IgG1
Immunogen Full length recombinant human Tubulin Beta 3 expressed in &italic_begin;E. coli&italic_end;. [UniProt No. Q13509].
Antigen beta-III Tubulin
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP142568SS

 
novus biologicals canada
NBP142568SS
Unlabeled 0.025mL Each for $109.00

NBP142568

 
novus biologicals canada
NBP142568
Unlabeled 0.1mL Each for $309.05
Description & Specifications

Specifications

Clone 2.00E+09
Host Species Mouse
Isotype IgG1
Immunogen Full length recombinant human Tubulin Beta 3 expressed in &italic_begin;E. coli&italic_end;. [UniProt No. Q13509].
Antigen beta-III Tubulin
Monoclonal or Polyclonal Monoclonal
Format Ascites
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Simple Western
Applications Western Blotting
Gene Accession No. Q13509
Gene ID (Entrez) 10381