Please login to your online account to display your discounted pricing

anti-beta-1,3-Glucuronyltransferase 1/B3GAT1/CD57, Clone: HNK-1, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP229405

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP229405

  • / Each
 

Description & Specifications

Specifications

Antigen 3-Glucuronyltransferase 1/B3GAT1/CD57
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Clone HNK-1
Conjugate Unlabeled
Cross Reactivity Human
Format Concentrated
Formulation Culture Medium
Gene Accession No. Q96E93
Host Species Mouse
Immunogen Membrane extracts of the HSB-2 T-lymphoblastoid line were used as the immunogen for the CD57 HNK-1 antibody.
Isotype κ
Quantity 0.5mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 27087
Test Specificity Anti-CD57 marks a subset of lymphocytes known as natural killer (NK) cells. Follicular center cell lymphomas often contain many NK cells within the neoplastic follicles. Anti-CD57 also stains neuroendocrine cells and their derived tumors, including carcinoid tumor and medulloblastoma. Anti-CD57 can also be useful in separating type B3 thymoma from thymic carcinoma when combined with a panel that includes antibodies against GLUT1, CD5, and CEA.