Please login to your online account to display your discounted pricing

anti-beta-1,3-Glucuronyltransferase 1/B3GAT1/CD57, Clone: HNK-1, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone HNK-1
Host Species Mouse
Isotype κ
Immunogen Membrane extracts of the HSB-2 T-lymphoblastoid line were used as the immunogen for the CD57 HNK-1 antibody.
Antigen 3-Glucuronyltransferase 1/B3GAT1/CD57
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP2294051M

 
novus biologicals canada
NBP2294051ML
Unlabeled 1mL Each for $429.00

NBP229405

 
novus biologicals canada
NBP229405
Unlabeled 0.5mL Each for $321.00
Description & Specifications

Specifications

Clone HNK-1
Host Species Mouse
Isotype κ
Immunogen Membrane extracts of the HSB-2 T-lymphoblastoid line were used as the immunogen for the CD57 HNK-1 antibody.
Antigen 3-Glucuronyltransferase 1/B3GAT1/CD57
Monoclonal or Polyclonal Monoclonal
Format Concentrated
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity Anti-CD57 marks a subset of lymphocytes known as natural killer (NK) cells. Follicular center cell lymphomas often contain many NK cells within the neoplastic follicles. Anti-CD57 also stains neuroendocrine cells and their derived tumors, including carcinoid tumor and medulloblastoma. Anti-CD57 can also be useful in separating type B3 thymoma from thymic carcinoma when combined with a panel that includes antibodies against GLUT1, CD5, and CEA.
Gene Accession No. Q96E93
Gene ID (Entrez) 27087
Formulation Culture Medium