Please login to your online account to display your discounted pricing

anti-Beclin 1/ATG6, Clone: 9B6, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP100088SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP100088SS

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Bovine
Cross Reactivity Chicken
Cross Reactivity Equine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Primate
Cross Reactivity Rat
Gene Accession No. Q14457
Regulatory Status RUO
Format Ascites
Primary or Secondary Primary
Conjugate Unlabeled
Monoclonal or Polyclonal Monoclonal
Immunogen Full length human Beclin 1 protein [Swiss-Prot No. Q14457]
Quantity 0.025mL
Applications Immunohistochemistry
Applications Western Blotting
Gene ID (Entrez) 8678
Antigen Beclin 1/ATG6
Clone 9B6
Test Specificity Human, mouse, rat, monkey, bovine, porcine, horse and chicken.
Host Species Mouse