Please login to your online account to display your discounted pricing

anti-Beclin 1/ATG6, Clone: 9B6, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP100088

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP100088

  • / Each
 

Description & Specifications

Specifications

Immunogen Full length human Beclin 1 protein [Swiss-Prot No. Q14457]
Cross Reactivity Bovine
Cross Reactivity Chicken
Cross Reactivity Equine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Primate
Cross Reactivity Rat
Quantity 0.1mL
Clone 9B6
Applications Immunohistochemistry
Applications Western Blotting
Format Ascites
Antigen Beclin 1/ATG6
Gene Accession No. Q14457
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 8678
Conjugate Unlabeled
Regulatory Status RUO
Test Specificity Human, mouse, rat, monkey, bovine, porcine, horse and chicken.
Host Species Mouse