Please login to your online account to display your discounted pricing

anti-Beclin 1/ATG6, Clone: 9B6, Novus Biologicals™

$139.00 - $309.05

Specifications

Clone 9B6
Host Species Murine
Immunogen Full length human Beclin 1 protein [Swiss-Prot No. Q14457]
Antigen Beclin 1/ATG6
Monoclonal or Polyclonal Monoclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP100088SS

 
novus biologicals canada
NBP100088SS
Unlabeled 0.025mL Each for $139.00

NBP100088

 
novus biologicals canada
NBP100088
Unlabeled 0.1mL Each for $309.05
Description & Specifications

Specifications

Clone 9B6
Host Species Murine
Immunogen Full length human Beclin 1 protein [Swiss-Prot No. Q14457]
Antigen Beclin 1/ATG6
Monoclonal or Polyclonal Monoclonal
Format Ascites
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Western Blot
Test Specificity Human, mouse, rat, monkey, bovine, porcine, horse and chicken.
Gene Accession No. Q14457
Gene ID (Entrez) 8678