Please login to your online account to display your discounted pricing

anti-Beclin 1/ATG6, Clone: 4H10, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP100085

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP100085

  • / Each
 

Description & Specifications

Specifications

Test Specificity Human. 99% sequence identity with monkey, 98% identity with mouse and bovine, 97% identity with porcine, rat and horse, 91% identity with chicken.
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Full length human Beclin 1 protein [Swiss-Prot No. Q14457]
Quantity 0.1mL
Clone 4H10
Concentration 1.0mg/mL
Formulation Tris-glycine, 150mM NaCl
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Conjugate Unlabeled
Isotype κ
Gene ID (Entrez) 8678
Cross Reactivity Human
Host Species Mouse
Format Purified
Antigen Beclin 1/ATG6
Regulatory Status RUO
Gene Accession No. Q14457