Please login to your online account to display your discounted pricing

anti-Beclin 1/ATG6, Clone: 4H10, Novus Biologicals™

$139.00 - $329.00

Specifications

Clone 4H10
Host Species Murine
Isotype κ
Immunogen Full length human Beclin 1 protein [Swiss-Prot No. Q14457]
Antigen Beclin 1/ATG6
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP100085SS

 
novus biologicals canada
NBP100085SS
Unlabeled 0.025mL Each for $139.00

NBP100085

 
novus biologicals canada
NBP100085
Unlabeled 0.1mL Each for $329.00
Description & Specifications

Specifications

Clone 4H10
Host Species Murine
Isotype κ
Immunogen Full length human Beclin 1 protein [Swiss-Prot No. Q14457]
Antigen Beclin 1/ATG6
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Test Specificity Human. 99% sequence identity with monkey, 98% identity with mouse and bovine, 97% identity with porcine, rat and horse, 91% identity with chicken.
Gene Accession No. Q14457
Gene ID (Entrez) 8678
Formulation Tris-glycine, 150mM NaCl
Concentration 1.0mg/mL