Please login to your online account to display your discounted pricing

anti-Beclin 1/ATG6, Clone: 1B7, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP100084

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP100084

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Monoclonal
Concentration 1mg/mL
Gene ID (Entrez) 8678
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Cross Reactivity Bovine
Cross Reactivity Chicken
Cross Reactivity Equine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Primate
Cross Reactivity Rat
Isotype κ
Host Species Mouse
Primary or Secondary Primary
Clone 1B7
Conjugate Unlabeled
Gene Accession No. Q14457
Formulation Tris-glycine, 150mM NaCl
Test Specificity Human, mouse, rat, monkey, bovine, porcine, horse and chicken.
Immunogen Full length human Beclin 1 protein [Swiss-Prot No. Q14457]
Quantity 0.1mL
Regulatory Status RUO
Antigen Beclin 1/ATG6
Format Purified