Please login to your online account to display your discounted pricing

anti-Beclin 1/ATG6, Clone: 1B7, Novus Biologicals™

$354.00 - $354.00

Specifications

Clone 1B7
Host Species Murine
Isotype κ
Immunogen Full length human Beclin 1 protein [Swiss-Prot No. Q14457]
Antigen Beclin 1/ATG6
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP100084

 
novus biologicals canada
NBP100084
Unlabeled 0.1mL Each for $354.00
Description & Specifications

Specifications

Clone 1B7
Host Species Murine
Isotype κ
Immunogen Full length human Beclin 1 protein [Swiss-Prot No. Q14457]
Antigen Beclin 1/ATG6
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Test Specificity Human, mouse, rat, monkey, bovine, porcine, horse and chicken.
Gene Accession No. Q14457
Gene ID (Entrez) 8678
Formulation Tris-glycine, 150mM NaCl
Concentration 1mg/mL