Please login to your online account to display your discounted pricing

anti-Beclin 1/ATG6, Clone: 1B7, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP100084SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP100084SS

  • / Each
 

Description & Specifications

Specifications

Host Species Mouse
Cross Reactivity Bovine
Cross Reactivity Chicken
Cross Reactivity Equine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Primate
Cross Reactivity Rat
Immunogen Full length human Beclin 1 protein [Swiss-Prot No. Q14457]
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Regulatory Status RUO
Clone 1B7
Format Purified
Formulation Tris-glycine, 150mM NaCl
Gene Accession No. Q14457
Primary or Secondary Primary
Test Specificity Human, mouse, rat, monkey, bovine, porcine, horse and chicken.
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 8678
Concentration 1mg/mL
Conjugate Unlabeled
Isotype κ
Quantity 0.025mL
Antigen Beclin 1/ATG6