Please login to your online account to display your discounted pricing

anti-Bcl-xL, Clone: 2H12, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP2329171ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP2329171M

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Rat
Antigen Bcl-xL
Immunogen A synthetic peptide
Quantity 1mL
Format Purified
Host Species Mouse
Clone 2H12
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Gene Accession No. Q07817
Conjugate Unlabeled
Gene ID (Entrez) 598
Formulation PBS with 0.05% BSA.
Primary or Secondary Primary
Concentration 0.2mg/mL
Isotype IgG2a