Please login to your online account to display your discounted pricing

anti-Bcl-xL, Clone: 2H12, Novus Biologicals™

$429.00 - $429.00

Specifications

Clone 2H12
Host Species Mouse
Isotype IgG2a
Immunogen A synthetic peptide
Antigen Bcl-xL
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP2329171M

 
novus biologicals canada
NBP2329171ML
Unlabeled 1mL Each for $429.00
Description & Specifications

Specifications

Clone 2H12
Host Species Mouse
Isotype IgG2a
Immunogen A synthetic peptide
Antigen Bcl-xL
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Gene Accession No. Q07817
Gene ID (Entrez) 598
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL