Please login to your online account to display your discounted pricing

anti-Bax inhibitor 1, Clone: 20F565, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224912

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224912

  • / Each
 

Description & Specifications

Specifications

Antigen Bax inhibitor 1
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Clone 20F565
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Purified
Formulation 100μg of antibody in 200μL PBS containing 0.05% BSA
Gene Accession No. P55061
Host Species Murine
Immunogen A mixture of three synthetic peptides corresponding to the N-terminal
Isotype κ
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 7009
Test Specificity This antibody is reactive to Human, Mouse, Rat.
Target Species Human
Target Species Murine
Target Species Rat