Please login to your online account to display your discounted pricing

anti-Bax inhibitor 1, Clone: 20F248, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP2-24949SS

 View all options for this product


For Research Use Only

Catalog No. NBP224949SS


Description & Specifications


Antigen Bax inhibitor 1
Applications Western Blotting
Clone 20F248
Conjugate Unlabeled
Cross Reactivity Human
Format Ascites
Formulation Ascited diluted with PBS
Gene Accession No. P55061
Host Species Mouse
Immunogen A mixture of three synthetic peptides corresponding to the N-terminal
Isotype IgM
Quantity 0.05mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 7009
Test Specificity This antibody is reactive to Human.