Please login to your online account to display your discounted pricing

anti-Bax inhibitor 1, Clone: 20F248, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224949

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224949

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. P55061
Primary or Secondary Primary
Gene ID (Entrez) 7009
Test Specificity This antibody is reactive to Human.
Immunogen A mixture of three synthetic peptides corresponding to the N-terminal
Isotype IgM
Conjugate Unlabeled
Quantity 0.2mL
Cross Reactivity Human
Format Ascites
Regulatory Status RUO
Host Species Mouse
Antigen Bax inhibitor 1
Applications Western Blotting
Formulation Ascited diluted with PBS
Clone 20F248
Monoclonal or Polyclonal Monoclonal