Please login to your online account to display your discounted pricing

anti-Bax inhibitor 1, Clone: 20F248, Novus Biologicals™

$309.05 - $309.05

Specifications

Clone 20F248
Host Species Mouse
Isotype IgM
Immunogen A mixture of three synthetic peptides corresponding to the N-terminal
Antigen Bax inhibitor 1
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224949

 
novus biologicals canada
NBP224949
Unlabeled 0.2mL Each for $309.05
Description & Specifications

Specifications

Clone 20F248
Host Species Mouse
Isotype IgM
Immunogen A mixture of three synthetic peptides corresponding to the N-terminal
Antigen Bax inhibitor 1
Monoclonal or Polyclonal Monoclonal
Format Ascites
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity This antibody is reactive to Human.
Gene Accession No. P55061
Gene ID (Entrez) 7009
Formulation Ascited diluted with PBS