Please login to your online account to display your discounted pricing

anti-B7-1/CD80, Clone: 62N3G8, Novus Biologicals™

$109.00 - $329.00

Specifications

Clone 62N3G8
Host Species Mouse
Isotype κ
Immunogen Amino acids 102-223 of human CD80 were used as the immunogen for the antibody.
Antigen B7-1/CD80
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP225255

 
novus biologicals canada
NBP225255
Unlabeled 0.1mg Each for $329.00

NBP225255SS

 
novus biologicals canada
NBP225255SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Clone 62N3G8
Host Species Mouse
Isotype κ
Immunogen Amino acids 102-223 of human CD80 were used as the immunogen for the antibody.
Antigen B7-1/CD80
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity This product reacts with Human.
Gene Accession No. NP_005182
Gene ID (Entrez) 941
Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA
Concentration 0.5mg/mL