Please login to your online account to display your discounted pricing

anti-Aurora A, Clone: 1F8, Novus Biologicals™

$109.00 - $309.05

Specifications

Clone 1F8
Host Species Murine
Isotype IgG1
Immunogen Purified recombinant fragment of human Aurora A expressed in &italic_begin;E. coli&italic_end;. [UniProt No. O14965]
Antigen Aurora A
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP222118

 
novus biologicals canada
NBP222118
Unlabeled 0.1mL Each for $309.05

NBP222118SS

 
novus biologicals canada
NBP222118SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Clone 1F8
Host Species Murine
Isotype IgG1
Immunogen Purified recombinant fragment of human Aurora A expressed in &italic_begin;E. coli&italic_end;. [UniProt No. O14965]
Antigen Aurora A
Monoclonal or Polyclonal Monoclonal
Format Precipitation
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blot
Gene Accession No. O14965
Gene ID (Entrez) 6790
Formulation PBS
Concentration 1.0mg/mL