Please login to your online account to display your discounted pricing

anti-ATF2 (p Thr71), Clone: 103C411.2, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056594SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056594T

  • / Each
 

Description & Specifications

Specifications

Antigen ATF2 (p Thr71)
Applications Flow Cytometry
Applications Western Blot
Clone 103C411.2
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS containing 0.05% BSA
Gene Accession No. P15336
Host Species Murine
Immunogen This antibody was generated by immunizing mice with a synthetic peptide containing phosphorylated threonine at position 71 of human ATF-2.
Isotype IgG1
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 1386
Test Specificity This monoclonal antibody is specific for phospho (Thr71) ATF2. It cannot be used to test for non-phosphorylated ATF2.
Target Species Human