Please login to your online account to display your discounted pricing

anti-ATF2 (p Thr71), Clone: 103C411.2, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056594

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056594

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. P15336
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 1386
Test Specificity This monoclonal antibody is specific for phospho (Thr71) ATF2. It cannot be used to test for non-phosphorylated ATF2.
Formulation PBS containing 0.05% BSA
Primary or Secondary Primary
Concentration 0.5mg/mL
Conjugate Unlabeled
Host Species Mouse
Applications Flow Cytometry
Applications Western Blotting
Immunogen This antibody was generated by immunizing mice with a synthetic peptide containing phosphorylated threonine at position 71 of human ATF-2.
Cross Reactivity Human
Isotype IgG1
Quantity 0.1mg
Format Purified
Antigen ATF2 (p Thr71)
Regulatory Status RUO
Clone 103C411.2