Please login to your online account to display your discounted pricing

anti-ARNT/HIF-1 beta, Clone: H1beta234, Novus Biologicals™


Manufacturer: novus biologicals canada  NB100124SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB100124SS

  • / Each
 

Description & Specifications

Specifications

Antigen ARNT/HIF-1 beta
Applications Chromatin Immunoprecipitation
Applications Gel Supershift
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Clone H1beta234
Concentration 1.4mg/mL
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Ferret
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Cross Reactivity Sheep
Format Purified
Formulation 1X PBS
Gene Accession No. P27540
Host Species Murine
Immunogen Fusion protein containing amino acids 496-789 of human HIF-1 beta. [UniProt No. P27540]
Isotype κ
Quantity 0.025mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 405
Target Species Bovine
Target Species Ferret
Target Species Human
Target Species Murine
Target Species Primate
Target Species Rat
Target Species Sheep