Please login to your online account to display your discounted pricing

anti-Apolipoprotein E/ApoE, Clone: 1H4, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP222110SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP222110SS

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Conjugate Unlabeled
Gene ID (Entrez) 348
Cross Reactivity Human
Clone 1H4
Immunogen Purified recombinant fragment of human ApoE expressed in &italic_begin;E. coli&italic_end;. [UniProt No. P02649]
Isotype IgG1
Regulatory Status RUO
Quantity 0.025mL
Format Ascites
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Antigen Apolipoprotein E/ApoE
Monoclonal or Polyclonal Monoclonal
Gene Accession No. P02649