Please login to your online account to display your discounted pricing

anti-Apolipoprotein E/ApoE, Clone: 1H4, Novus Biologicals™

$109.00 - $309.05

Specifications

Clone 1H4
Host Species Murine
Isotype IgG1
Immunogen Purified recombinant fragment of human ApoE expressed in &italic_begin;E. coli&italic_end;. [UniProt No. P02649]
Antigen Apolipoprotein E/ApoE
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP222110

 
novus biologicals canada
NBP222110
Unlabeled 0.1mL Each for $309.05

NBP222110SS

 
novus biologicals canada
NBP222110SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Clone 1H4
Host Species Murine
Isotype IgG1
Immunogen Purified recombinant fragment of human ApoE expressed in &italic_begin;E. coli&italic_end;. [UniProt No. P02649]
Antigen Apolipoprotein E/ApoE
Monoclonal or Polyclonal Monoclonal
Format Ascites
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Gene Accession No. P02649
Gene ID (Entrez) 348