Please login to your online account to display your discounted pricing

anti-Apolipoprotein A5, Clone: 1G5G9, Novus Biologicals™

$109.00 - $309.05

Specifications

Clone 1G5G9
Host Species Murine
Isotype κ
Immunogen Ni-NTA purified full-length recombinant human Apolipoprotein A5 expressed in &italic_begin;E. coli&italic_end; strain BL21 (DE3). [UniProt No. Q6Q788]
Antigen Apolipoprotein A5
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB11055454

 
novus biologicals canada
NB11055454
Unlabeled 0.1mL Each for $309.05

NB11055454T

 
novus biologicals canada
NB11055454SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Clone 1G5G9
Host Species Murine
Isotype κ
Immunogen Ni-NTA purified full-length recombinant human Apolipoprotein A5 expressed in &italic_begin;E. coli&italic_end; strain BL21 (DE3). [UniProt No. Q6Q788]
Antigen Apolipoprotein A5
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Gene Accession No. Q6Q788
Gene ID (Entrez) 116519
Formulation PBS, pH 7.4
Concentration 1.9mg/mL