Please login to your online account to display your discounted pricing

anti-APIP, Clone: 19F424, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056568SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056568T

  • / Each
 

Description & Specifications

Specifications

Concentration 0.5mg/mL
Monoclonal or Polyclonal Monoclonal
Test Specificity This monoclonal antibody can be used for detection of APIP and APIP2.
Primary or Secondary Primary
Gene ID (Entrez) 51074
Gene Accession No. Q96GX9
Formulation PBS containing 0.2% gelatin
Conjugate Unlabeled
Cross Reactivity Human
Host Species Mouse
Quantity 0.025mg
Regulatory Status RUO
Applications Western Blotting
Clone 19F424
Isotype κ
Immunogen The antibody was developed against a full-length His-tagged recombinant APIP protein.
Format Purified
Antigen APIP