Please login to your online account to display your discounted pricing

anti-APIP, Clone: 19F424, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056568

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056568

  • / Each
 

Description & Specifications

Specifications

Antigen APIP
Formulation PBS containing 0.2% gelatin
Gene Accession No. Q96GX9
Isotype κ
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 51074
Test Specificity This monoclonal antibody can be used for detection of APIP and APIP2.
Concentration 0.5mg/mL
Conjugate Unlabeled
Regulatory Status RUO
Cross Reactivity Human
Applications Western Blotting
Immunogen The antibody was developed against a full-length His-tagged recombinant APIP protein.
Format Purified
Host Species Mouse
Quantity 0.1mg
Clone 19F424