Please login to your online account to display your discounted pricing

anti-APIP, Clone: 19F424, Novus Biologicals™

$139.00 - $329.00

Specifications

Clone 19F424
Host Species Mouse
Isotype κ
Immunogen The antibody was developed against a full-length His-tagged recombinant APIP protein.
Antigen APIP
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056568

 
novus biologicals canada
NB10056568
Unlabeled 0.1mg Each for $329.00

NB10056568T

 
novus biologicals canada
NB10056568SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Clone 19F424
Host Species Mouse
Isotype κ
Immunogen The antibody was developed against a full-length His-tagged recombinant APIP protein.
Antigen APIP
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity This monoclonal antibody can be used for detection of APIP and APIP2.
Gene Accession No. Q96GX9
Gene ID (Entrez) 51074
Formulation PBS containing 0.2% gelatin
Concentration 0.5mg/mL