Please login to your online account to display your discounted pricing

anti-APE, Clone: 13B8E5C2, Novus Biologicals™

$168.10 - $384.00

Specifications

Clone 13B8E5C2
Host Species Murine
Isotype IgG2b
Immunogen Purified human APE1 [UniProt No. P27695]
Antigen APE
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB100116SS

 
novus biologicals canada
NB100116SS
Unlabeled 0.025mL Each for $168.10

NB100116

 
novus biologicals canada
NB100116
Unlabeled 0.05mL Each for $384.00
Description & Specifications

Specifications

Clone 13B8E5C2
Host Species Murine
Isotype IgG2b
Immunogen Purified human APE1 [UniProt No. P27695]
Antigen APE
Monoclonal or Polyclonal Monoclonal
Format Ascites
Primary or Secondary Primary
Regulatory Status RUO
Applications Chromatin Immunoprecipitation
Applications Gel Supershift
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Simple Western
Applications Western Blot
Gene Accession No. P27695
Gene ID (Entrez) 328
Formulation PBS